5-Aminomethyl-2-(boc-amino)pyridine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Amino-5-(aminomethyl)pyridine, 2-BOC protected, 2-(Boc-amino)-5-(aminomethyl)pyridine
H50078.03 H50078.06
H50078.03EA 5200 SEK
H50078.03 H50078.06
5-Aminomethyl-2-(boc-amino)pyridine 97%
5-Aminomethyl-2-(boc-amino)pyridine
Purity: 97% CAS No.: 187237-37-2 Molecular Formula: C11H17N3O2 Formula Weight: 223.28 Melting Point: 132-134° MDL No.: MFCD07781132
Formel: C₁₁H₁₇N₃O₂
MW: 223,28 g/mol
Smältpunkt: 132…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD07781132
CAS nummer: 187237-37-2