Piperazin hexahydrat 97+%
Leverantör: Thermo Scientific
Fara
Synonyms: Piperazine hydrate
H37069.0B H37069.30
H37069.0BEA 2675 SEK
H37069.0B H37069.30
Piperazin hexahydrat 97+%
Piperazin hexahydrat
Purity: 97+% CAS No.: 142-63-2 Molecular Formula: C4H10N2.xH2O Formula Weight: 86.14anhy Flash Point: 100°(212°F) Physical Form: Crystalline powder or flake Appearance: White Odour: Weak, characteristic pH: 11-12 Melting Point: 42-46° Boiling Point: 125-130° UN No.: UN2579 MDL No.: MFCD00149389
Formel: C₄H₁₀N₂·6H₂O
MW: 194,23 g/mol
Kokpunkt: 125…130 °C
Smältpunkt: 42…46 °C
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00149389
CAS nummer: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III