3-(2-Methoxyphenyl)-1H-pyrazole 97%
Leverantör: Thermo Scientific
Varning
H34492.03 H34492.06
H34492.03EA 1925 SEK
H34492.03 H34492.06
3-(2-Methoxyphenyl)-1H-pyrazole 97%
3-(2-Methoxyphenyl)-1H-pyrazole
Purity: 97% CAS No.: 59843-63-9 Molecular Formula: C10H10N2O Formula Weight: 174.20 Melting Point: 82-84° MDL No.: MFCD02091524
Formel: C₁₀H₁₀N₂O
MW: 174,20 g/mol
Smältpunkt: 82…84 °C
Storage Temperature: Omgivande
MDL Number: MFCD02091524
CAS nummer: 59843-63-9