4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone
H34429.14 H34429.03 H34429.06
H34429.14EA 8175 SEK
H34429.14 H34429.03 H34429.06
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Purity: 97% CAS No.: 582-65-0 Molecular Formula: C10H6F4O2 Formula Weight: 234.15 Melting Point: 40-44° MDL No.: MFCD03419766
Formel: C₁₀H₆F₄O₂
MW: 234,15 g/mol
Smältpunkt: 40…44 °C
Storage Temperature: Omgivande
MDL Number: MFCD03419766
CAS nummer: 582-65-0