Furo[3,2-b]pyridine-2-carboxylic acid 97%
Leverantör: Thermo Scientific
Varning
H34400.MD H34400.03 H34400.06
H34400.MDEA 1275 SEK
H34400.MD H34400.03 H34400.06
Furo[3,2-b]pyridine-2-carboxylic acid 97%
Furo[3,2-b]pyridine-2-carboxylic acid
Purity: 97% CAS No.: 112372-14-2 Molecular Formula: C8H5NO3 Formula Weight: 163.13 Melting Point: 300° MDL No.: MFCD11588128
Formel: C₈H₅NO₃
MW: 163,13 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD11588128
CAS nummer: 112372-14-2