trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: trans-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
H34384.14 H34384.22
H34384.14EA 2225 SEK
H34384.14 H34384.22
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Purity: 98% CAS No.: 49708-81-8 Molecular Formula: C13H15ClO2 Formula Weight: 238.71 Melting Point: 249-254° MDL No.: MFCD06797626
Formel: C₁₃H₁₅ClO₂
MW: 238,71 g/mol
Smältpunkt: 249…254 °C
Storage Temperature: Omgivande
MDL Number: MFCD06797626
CAS nummer: 49708-81-8