1-Methylindole-3-carbonitrile 96%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Methyl-1H-indole-3-carbonitrile
H34157.MD H34157.03 H34157.06
H34157.MDEA 795 SEK
H34157.MD H34157.03 H34157.06
1-Methylindole-3-carbonitrile 96%
1-Methylindole-3-carbonitrile
Purity: 96% CAS No.: 24662-37-1 Molecular Formula: C10H8N2 Formula Weight: 156.19 Melting Point: 55-56° MDL No.: MFCD00466602
Formel: C₁₀H₈N₂
MW: 156,19 g/mol
Smältpunkt: 55…56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00466602
CAS nummer: 24662-37-1