3-(4,4-Dimethyl-2-oxazolinyl)pyridine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(4,5-Dihydro-4,4-dimethyl-1,3-oxazol-2-yl)pyridine, 4,4-Dimethyl-2-(pyridin-3-yl)-4,5-dihydrooxazole
H33997.09 H33997.03
H33997.09EA 5425 SEK
H33997.09 H33997.03
3-(4,4-Dimethyl-2-oxazolinyl)pyridine 97%
3-(4,4-Dimethyl-2-oxazolinyl)pyridine
Purity: 97% CAS No.: 68981-86-2 Molecular Formula: C10H12N2O Formula Weight: 176.22 Melting Point: 25-27° MDL No.: MFCD01313220
Formel: C₁₀H₁₂N₂O
MW: 176,22 g/mol
Smältpunkt: 25…27 °C
Storage Temperature: Omgivande
MDL Number: MFCD01313220
CAS nummer: 68981-86-2