2-(2-Phthalimidoethoxy)acetic acid 97%
Leverantör: Thermo Fisher Scientific
H33988.03 H33988.06
H33988.03EA 1275 SEK
H33988.03 H33988.06
2-(2-Phthalimidoethoxy)acetic acid 97%
2-(2-Phthalimidoethoxy)acetic acid
Purity: 97% CAS No.: 69676-65-9 Molecular Formula: C12H11NO5 Formula Weight: 249.22 Melting Point: 130-132° MDL No.: MFCD06661364
Formel: C₁₂H₁₁NO₅
Smältpunkt: 130…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD06661364
CAS nummer: 69676-65-9