3-(1-Naphthyl)propionic acid 96%
Leverantör: Thermo Scientific
Varning
H33795.03 H33795.06
H33795.03EA 1775 SEK
H33795.03 H33795.06
3-(1-Naphthyl)propionic acid 96%
3-(1-Naphthyl)propionic acid
Purity: 96% CAS No.: 3243-42-3 Molecular Formula: C13H12O2 Formula Weight: 200.24 Melting Point: 155-159° MDL No.: MFCD00033058
Formel: C₁₃H₁₂O₂
MW: 200,24 g/mol
Smältpunkt: 155…159 °C
Storage Temperature: Omgivande
MDL Number: MFCD00033058
CAS nummer: 3243-42-3