(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Difluoro-1-methylcyclopropanecarboxylic acid
H33055.MD H33055.03
H33055.MDEA 2300 SEK
H33055.MD H33055.03
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
Purity: 97% CAS No.: 128073-33-6 Molecular Formula: C5H6F2O2 Formula Weight: 136.10 Melting Point: 62-64° MDL No.: MFCD07777163
Formel: C₅H₆F₂O₂
MW: 136,10 g/mol
Smältpunkt: 62…64 °C
Storage Temperature: Omgivande
MDL Number: MFCD07777163
CAS nummer: 128073-33-6