5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
Leverantör: Thermo Fisher Scientific
Varning
H33044.03 H33044.06
H33044.03EA 2075 SEK
H33044.03 H33044.06
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride
Purity: 96% CAS No.: 84163-16-6 Molecular Formula: C12H13FN2O.HCl Formula Weight: 256.71 Melting Point: ca 298° dec. MDL No.: MFCD06200879
Formel: C₁₂H₁₄ClFN₂O
MW: 256.71 g/mol
Smältpunkt: ∼ 298 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD06200879
CAS nummer: 84163-16-6