2-Methyl-1,3-benzothiazol-5-ol 97%
Leverantör: Thermo Fisher Scientific
Varning
H32937.03 H32937.09
H32937.03EA 855 SEK
H32937.03 H32937.09
2-Methyl-1,3-benzothiazol-5-ol 97%
2-Methyl-1,3-benzothiazol-5-ol
Purity: 97% CAS No.: 68867-14-1 Molecular Formula: C8H7NOS Formula Weight: 165.21 Melting Point: 185-187° MDL No.: MFCD00192276
Formel: C₈H₇NOS
MW: 165.22 g/mol
Smältpunkt: 185…187 °C
Storage Temperature: Omgivande
MDL Number: MFCD00192276
CAS nummer: 68867-14-1