4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene
H32695.06 H32695.14
H32695.06EA 1550 SEK
H32695.06 H32695.14
4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%
4,5-Dimethyl-o-phenylenediamine
Purity: 98% CAS No.: 3171-45-7 Molecular Formula: C8H12N2 Formula Weight: 136.20 Storage Temperature: 2°C to 8°C Melting Point: 127-129° UN No.: UN2811 MDL No.: MFCD00007729
Formel: C₈H₁₂N₂
MW: 136.2 g/mol
Smältpunkt: 127…129 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007729
CAS nummer: 3171-45-7
EINECS: 221-635-1
UN: 2811
ADR: 6.1 III