1-(4-Chlorophenyl)-1-methylethylamine 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(4-Chlorophenyl)propan-2-amine
H32527.MD H32527.03
H32527.MDEA 2300 SEK
H32527.MD H32527.03
1-(4-Chlorophenyl)-1-methylethylamine 97%
1-(4-Chlorophenyl)-1-methylethylamine
Purity: 97% CAS No.: 17797-11-4 Molecular Formula: C9H12ClN Formula Weight: 169.66 Refractive Index: 1.5330 Odour: Amine-like Boiling Point: 43-45°/0.3mm UN No.: UN2735 MDL No.: MFCD08669636
Formel: C₉H₁₂ClN
MW: 169,66 g/mol
Kokpunkt: 43…45 °C (0,3 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD08669636
CAS nummer: 17797-11-4
ADR: 8,III