2-Ethyl-5,5-dimethyl-1,3-dioxane 99%
Leverantör: Thermo Scientific
Varning
H32351.06 H32351.14
H32351.06EA 2100 SEK
H32351.06 H32351.14
2-Ethyl-5,5-dimethyl-1,3-dioxane 99%
2-Ethyl-5,5-dimethyl-1,3-dioxane
Purity: 99% CAS No.: 768-58-1 Molecular Formula: C8H16O2 Formula Weight: 144.21 Flash Point: 50°(122°F) Odour: Characteristic Boiling Point: 154° UN No.: UN1993 MDL No.: MFCD16622268
Formel: C₈H₁₆O₂
Kokpunkt: 154 °C
Densitet: 0,88
Flampunkt: 50 °C (122 °F)
Storage Temperature: Omgivande
MDL Number: MFCD16622268
CAS nummer: 768-58-1
ADR: 3,III