2-Ethyl-4-methyl-1,3-dioxolane cis- and trans-mixture 99%
Leverantör: Thermo Fisher Scientific
Varning
H32187.06 H32187.14
H32187.06EA 2325 SEK
H32187.06 H32187.14
2-Ethyl-4-methyl-1,3-dioxolane cis- and trans-mixture 99%
2-Ethyl-4-methyl-1,3-dioxolane
Purity: 99% CAS No.: 4359-46-0 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 29°(84°F) Refractive Index: 1.4030 Odour: Characteristic Boiling Point: 117° UN No.: UN1993 MDL No.: MFCD16622267
Formel: C₆H₁₂O₂
Kokpunkt: 117 °C
Densitet: 0,904
Flampunkt: 29 °C (84 °F)
Storage Temperature: Omgivande
MDL Number: MFCD16622267
CAS nummer: 4359-46-0
UN: 1993
ADR: 3,III