11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide 95%
Leverantör: Thermo Scientific
Varning
Synonyms: IR-780, 2-(2-(2-Chloro-3-(2-(3,3-dimethyl-1-propylindolin-2-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-3,3-dimethyl-1-propyl-3H-indol-1-ium iodide
H32150.03 H32150.06
H32150.03EA 2800 SEK
H32150.03 H32150.06
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide 95%
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide
Purity: 95% CAS No.: 207399-07-3 Molecular Formula: C36H44ClIN2 Formula Weight: 667.12 Melting Point: 232-234° MDL No.: MFCD00191907
Formel: C₃₆H₄₄ClIN₂
MW: 667,12 g/mol
Smältpunkt: 232…234 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191907
CAS nummer: 207399-07-3