Methyl-4,5-dibromo-2-thiophenecarboxylate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 4,5-dibromothiophene-2-carboxylate
H32113.03 H32113.06
H32113.03EA 1675 SEK
H32113.03 H32113.06
Methyl-4,5-dibromo-2-thiophenecarboxylate 97%
Methyl-4,5-dibromo-2-thiophenecarboxylate
Purity: 97% CAS No.: 62224-24-2 Molecular Formula: C6H4Br2O2S Formula Weight: 299.97 Melting Point: 77-81° MDL No.: MFCD00831688
Formel: C₆H₄Br₂O₂S
MW: 299,98 g/mol
Smältpunkt: 77…81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00831688
CAS nummer: 62224-24-2