Bromaminic acid ≥90%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonic acid
H32110.14 H32110.22
H32110.14EA 1975 SEK
H32110.14 H32110.22
Bromaminic acid ≥90%
Bromaminic acid
Purity: 90+% CAS No.: 116-81-4 Molecular Formula: C14H8BrNO5S Formula Weight: 382.19 Odour: Odourless Melting Point: ca 280° dec. MDL No.: MFCD00035694
Formel: C₁₄H₈BrNO₅S
MW: 382,19 g/mol
Smältpunkt: ∼ 280 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00035694
CAS nummer: 116-81-4
EINECS: 204-159-9