6-Bromothiochroman-4-one 95%
Leverantör: Thermo Fisher Scientific
Varning
H31968.03 H31968.06
H31968.03EA 1525 SEK
H31968.03 H31968.06
6-Bromothiochroman-4-one 95%
6-Bromothiochroman-4-one
Purity: 95% CAS No.: 13735-13-2 Molecular Formula: C9H7BrOS Formula Weight: 243.12 Physical Form: Solid Appearance: Pale yellow to pale brown Melting Point: 66-69° MDL No.: MFCD05664869
Formel: C₉H₇BrOS
MW: 243.12 g/mol
Smältpunkt: 66…69 °C
Storage Temperature: Omgivande
MDL Number: MFCD05664869
CAS nummer: 13735-13-2