4-Isopropenyltoluene stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 4,alpha-Dimethylstyrene, 1-Isopropenyl-4-methylbenzene
Stabilization: med 4-tert-Butylpyrokatekol
H31739.06 H31739.14 H31739.22
H31739.06EA 1350 SEK
H31739.06 H31739.14 H31739.22
4-Isopropenyltoluene stabilised
4-Isopropenyltoluene
Stab. with 4-tert-butylcatechol
CAS No.: 1195-32-0 Molecular Formula: C10H12 Formula Weight: 132.21 Flash Point: 63°(145°F) Refractive Index: 1.5090 Boiling Point: 186-189° MDL No.: MFCD00036510
Formel: C₁₀H₁₂
MW: 132,21 g/mol
Kokpunkt: 186…189 °C
Densitet: 0,872
Flampunkt: 63 °C (145 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00036510
CAS nummer: 1195-32-0
EINECS: 214-795-9