Thieno[3,2-b]thiophene 97%
Leverantör: Thermo Scientific
Synonyms: 1,4-Dithiapentalene
H30637.03 H30637.06
H30637.03EA 3150 SEK
H30637.03 H30637.06
Thieno[3,2-b]thiophene 97%
Thieno[3,2-b]thiophene
Purity: 97% CAS No.: 251-41-2 Molecular Formula: C6H4S2 Formula Weight: 140.23 Melting Point: 54-58° Boiling Point: 71°/1mm MDL No.: MFCD00179479
Formel: C₆H₄S₂
MW: 140,23 g/mol
Kokpunkt: 71 °C (1 mmHg)
Smältpunkt: 54…58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00179479
CAS nummer: 251-41-2