tert-Butyl-3-aminoazetidine-1-carboxylate 94%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 3-Amino-1-Boc-azetidine, tert-Butyl 3-aminoazetidine-1-carboxylate, 1-BOC-3-aminoazetidine
H28218.MD H28218.03
H28218.MDEA 2800 SEK
H28218.MD H28218.03
tert-Butyl-3-aminoazetidine-1-carboxylate 94%
tert-Butyl-3-aminoazetidine-1-carboxylate
Purity: 94% CAS No.: 193269-78-2 Molecular Formula: C8H16N2O2 Formula Weight: 172.23 Refractive Index: 1.4650 UN No.: UN2735 MDL No.: MFCD01861753
Formel: C₈H₁₆N₂O₂
MW: 172.23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01861753
CAS nummer: 193269-78-2
ADR: 8 III