N-Boc-cis-4-hydroxy-L-proline 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-BOC-cis-4-Hydroxy-L-proline, cis-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-cis-Hyp-OH, cis-N-Boc-4-hydroxy-L-proline, Boc-Cis-Hyp-OH, (2S,4S)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
H27850.03 H27850.06
H27850.03EA 2900 SEK
H27850.03 H27850.06
N-Boc-cis-4-hydroxy-L-proline 97%
N-Boc-cis-4-hydroxy-L-proline
Purity: 97% CAS No.: 87691-27-8 Molecular Formula: C10H17NO5 Formula Weight: 231.25 Melting Point: ca 146° dec. MDL No.: MFCD02094406
Formel: C₁₀H₁₇NO₅
MW: 231,248 g/mol
Smältpunkt: ∼ 146 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD02094406
CAS nummer: 87691-27-8
UN: 3077
ADR: 9,III