2-Methylcyclopropanecarboxylic acid cis- and trans-mixture 97%
Leverantör: Thermo Fisher Scientific
Fara
H27815.09 H27815.03
H27815.09EA 2450 SEK
H27815.09 H27815.03
2-Methylcyclopropanecarboxylic acid cis- and trans-mixture 97%
2-Methylcyclopropanecarboxylic acid
Purity: 97% CAS No.: 29555-02-0 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 88°(190°F) Refractive Index: 1.439 Boiling Point: 200-203° Beilstein Reference: 2323873 UN No.: UN3265 MDL No.: MFCD00001293
Formel: C₅H₈O₂
Kokpunkt: 200…203 °C
Densitet: 1,027
Flampunkt: 88 °C (190 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001293
CAS nummer: 29555-02-0
EINECS: 249-693-3
UN: 3265
ADR: 8,II