2-Cyclopentene-1-acetic acid 90% remainder mainly 3-cyclopentene-1-acetic acid, tech.
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(Cyclopent-2-en-1-yl)acetic acid
H27507.14 H27507.06
H27507.14EA 6550 SEK
H27507.14 H27507.06
2-Cyclopentene-1-acetic acid 90% remainder mainly 3-cyclopentene-1-acetic acid, tech.
2-Cyclopentene-1-acetic acid
Purity: 90% CAS No.: 13668-61-6 Molecular Formula: C7H10O2 Formula Weight: 126.15 Flash Point: 143°(289°F) Refractive Index: 1.4680 Odour: Rancid Melting Point: 19° Boiling Point: 238° Beilstein Reference: 2042172 MDL No.: MFCD00001400
Formel: C₇H₁₀O₂
MW: 126.15 g/mol
Kokpunkt: 238 °C
Smältpunkt: 19 °C
Densitet: 1.047
Flampunkt: 143 °C (289 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001400
CAS nummer: 13668-61-6
EINECS: 237-146-1