(S)-1-Cyclopropylethylamine ≥98%, ee 98%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (S)-1-Cyclopropylethanamine
H27499.06 H27499.03
H27499.06EA 5475 SEK
H27499.06 H27499.03
(S)-1-Cyclopropylethylamine ≥98%, ee 98%, ChiPros®
(S)-1-Cyclopropylethylamine
Purity: 98% CAS No.: 195604-39-8 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 3°(37°F) Melting Point: -68° Boiling Point: 95° UN No.: UN2733 MDL No.: MFCD08064289
Formel: C₅H₁₁N
MW: 85,15 g/mol
Kokpunkt: 95 °C
Smältpunkt: –68 °C
Densitet: 0,799
Flampunkt: 3 °C (37 °F)
Storage Temperature: Omgivande
MDL Number: MFCD08064289
CAS nummer: 195604-39-8
UN: 2733
ADR: 3,II