3-Bromomethylquinoxaline-2-one 90%, tech.
Leverantör: Thermo Fisher Scientific
Fara
H27169.14 H27169.06
H27169.14EA 2550 SEK
H27169.14 H27169.06
3-Bromomethylquinoxaline-2-one 90%, tech.
3-Bromomethylquinoxaline-2-one
Purity: 90% CAS No.: 62235-61-4 Molecular Formula: C9H7BrN2O Formula Weight: 239.07 Melting Point: 300° dec. UN No.: UN1759 MDL No.: MFCD02660613
Formel: C₉H₇BrN₂O
MW: 239.07 g/mol
Smältpunkt: >300 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD03412060
CAS nummer: 62235-61-4
UN: 1759
ADR: 8 III