Boc-trans-4-hydroxy-L-proline 97%
Leverantör: Thermo Fisher Scientific
Synonyms: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline
H27110.06 H27110.14
H27110.06EA 1425 SEK
H27110.06 H27110.14
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline
Purity: 97% CAS No.: 13726-69-7 Molecular Formula: C10H17NO5 Formula Weight: 231.25 Melting Point: 123-127° MDL No.: MFCD00053370
Formel: C₁₀H₁₇NO₅
MW: 231.3 g/mol
Smältpunkt: 123…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00053370
CAS nummer: 13726-69-7