(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
Leverantör: Thermo Scientific
Varning
Synonyms: Bis[(S)-1-phenylethyl]amine
H27106.14 H27106.06 H27106.03
H27106.14EA 12950 SEK
H27106.14 H27106.06 H27106.03
(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
(-)-Bis[(S)-1-phenylethyl]amine
Purity: 99% CAS No.: 56210-72-1 Molecular Formula: C16H19N Formula Weight: 225.34 Boiling Point: 98°/0.3mm MDL No.: MFCD00243087
Formel: C₁₆H₁₉N
MW: 225,34 g/mol
Kokpunkt: 98 °C (0,3 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00243087
CAS nummer: 56210-72-1