(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
(1S,2S)-(+)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-TsDPEN, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide
H27006.03
H27006.06
H27006.14
H27006.03EA
2950
SEK
H27006.03
H27006.06
H27006.14
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide
CAS No.: 167316-27-0
Molecular Formula: C21H22N2O2S
Formula Weight: 366.48
Melting Point: 127-130°
Optical Rotation: +29° (c=1 in chloroform)
MDL No.: MFCD03095684
Formel:
C₂₁H₂₂N₂O₂S Smältpunkt: 127…130 °C Storage Temperature: Omgivande |
MDL Number:
MFCD03095684 CAS nummer: 167316-27-0 |