2-(3,4-Dimethoxyphenyl)ethanol 98%
Leverantör: Thermo Scientific
Synonyms: Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol
H26941.18 H26941.09
H26941.18EA 7100 SEK
H26941.18 H26941.09
2-(3,4-Dimethoxyphenyl)ethanol 98%
2-(3,4-Dimethoxyphenyl)ethanol
Purity: 98% CAS No.: 7417-21-2 Molecular Formula: C10H14O3 Formula Weight: 182.22 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 172-174°/17mm MDL No.: MFCD00002894
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Kokpunkt: 172…174 °C (17 mmHg)
Smältpunkt: 45…49 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002894
CAS nummer: 7417-21-2
EINECS: 231-032-5