Cumylamine 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: alpha,alpha-Dimethylbenzylamine, Phenylisopropylamine, alpha-Aminoisopropylbenzene, 2-Phenylpropan-2-amine
H26845.03 H26845.06
H26845.03EA 3725 SEK
H26845.03 H26845.06
Cumylamine 96%
Cumylamine
Purity: 96% CAS No.: 585-32-0 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 72°(162°F) Refractive Index: 1.5200 Odour: Amine-like Boiling Point: 197° UN No.: UN2735 MDL No.: MFCD00134680
Formel: C₉H₁₃N
MW: 135.21 g/mol
Kokpunkt: 197 °C
Densitet: 0.94
Flampunkt: 72 °C (162 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00134680
CAS nummer: 585-32-0
EINECS: None
ADR: 8 III