(±)-Propranolol hydrochloride 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, Propranolol hydrochloride, DL-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride
H26645.14 H26645.06
H26645.14EA 5750 SEK
H26645.14 H26645.06
(±)-Propranolol hydrochloride 99%
(±)-Propranolol hydrochloride
Purity: 99% CAS No.: 318-98-9 Molecular Formula: C16H21NO2.HCl Formula Weight: 295.80 Melting Point: 163-165° MDL No.: MFCD00012558
Formel: C₁₆H₂₁NO₂·HCl
MW: 295,81 g/mol
Smältpunkt: 163…165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012558
CAS nummer: 318-98-9
EINECS: 206-268-7
Merck Index: 14,7840