4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate ≥97%
Leverantör: Thermo Scientific
Fara
H26333.03 H26333.06
H26333.03EA 1275 SEK
H26333.03 H26333.06
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate ≥97%
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate
Purity: 97+% CAS No.: 3945-69-5 Molecular Formula: C10H17ClN4O3.xH2O Formula Weight: 276.72(anhy) Melting Point: 110-114° Beilstein Reference: 8026872 UN No.: UN3261 MDL No.: MFCD03613550
Formel: C₁₀H₁₇ClN₄O₃·xH₂O
MW: 276,72 g/mol (anhy)
Smältpunkt: 110…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD03613550
CAS nummer: 3945-69-5
ADR: 8,III