2-(Trifluoromethyl)pyrrolidine 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Aza-2-(trifluoromethyl)cyclopentane, 2-(Trifluoromethyl)tetrahydropyrrole
H25852.MD H25852.03
H25852.MDEA 1875 SEK
H25852.MD H25852.03
2-(Trifluoromethyl)pyrrolidine 95%
2-(Trifluoromethyl)pyrrolidine
Purity: 95% CAS No.: 109074-67-1 Molecular Formula: C5H8F3N Formula Weight: 139.12 Flash Point: 26°(78°F) Refractive Index: 1.379 Boiling Point: 110-112° UN No.: UN2733 MDL No.: MFCD02663405
Formel: C₅H₈F₃N
MW: 139.12 g/mol
Kokpunkt: 110…112 °C
Densitet: 1.21
Flampunkt: 26 °C (78 °F)
Storage Temperature: Omgivande
MDL Number: MFCD02663405
CAS nummer: 109074-67-1
ADR: 3 III