Ethyl-4-hydroxycyclohexanecarboxylate cis- and trans-mixture 97%
Leverantör: Thermo Fisher Scientific
B25572.14 B25572.06
B25572.14EA 3975 SEK
B25572.14 B25572.06
Ethyl-4-hydroxycyclohexanecarboxylate cis- and trans-mixture 97%
Ethyl-4-hydroxycyclohexanecarboxylate
Purity: 97% CAS No.: 17159-80-7 Molecular Formula: C9H16O3 Formula Weight: 172.22 Flash Point: 110(230°F) Refractive Index: 1.4670 Boiling Point: 127-134°/0.1mm MDL No.: MFCD00192156
Formel: C₉H₁₆O₃
Kokpunkt: 127…134 °C (0,1 mmHg)
Densitet: 1,068
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00192156
CAS nummer: 17159-80-7
EINECS: 241-215-0