5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde
B24973.14 B24973.22
B24973.14EA 1675 SEK
B24973.14 B24973.22
5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
5-Norbornene-2-carboxaldehyde
Purity: 95% CAS No.: 5453-80-5 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 50°(122°F) Refractive Index: 1.4910 Odour: Characteristic Boiling Point: 68-70°/12mm Beilstein Reference: 1363813 UN No.: UN1989 MDL No.: MFCD00167568
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokpunkt: 68…70 °C (12 mmHg)
Densitet: 1,032
Flampunkt: 50 °C (122 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00167568
CAS nummer: 5453-80-5
EINECS: 226-698-9
UN: 1989
ADR: 3,III