Suberoyl chloride 97%
Leverantör: Thermo Fisher Scientific
Fara
B24580.06 B24580.14
B24580.06EA 2125 SEK
B24580.06 B24580.14
Suberoyl chloride 97%
Suberoyl chloride
Purity: 97% CAS No.: 10027-07-3 Molecular Formula: C8H12Cl2O2 Formula Weight: 211.09 Flash Point: 110°(230°F) Refractive Index: 1.4680 Boiling Point: 162-163°/15mm UN No.: UN3265 MDL No.: MFCD00000764
Formel: C₈H₁₂Cl₂O₂
Kokpunkt: 162…163 °C (15 mmHg)
Densitet: 1,172
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000764
CAS nummer: 10027-07-3
ADR: 8,II