Sodium 2,4-pentanedionate hydrate 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Sodium acetylacetonate hydrate
B24009.0B B24009.18 B24009.30
B24009.0BEA 11250 SEK
B24009.0B B24009.18 B24009.30
Sodium 2,4-pentanedionate hydrate 95%
Sodium 2,4-pentanedionate hydrate
Purity: 95% CAS No.: 86891-03-4 Molecular Formula: C5H7NaO2.xH2O Formula Weight: 122.11(anhy) Melting Point: 228-230° MDL No.: MFCD00078034
Formel: C₅H₇NaO₂·xH₂O
MW: 122,11 g/mol (anhy)
Smältpunkt: 228…230 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078034
CAS nummer: 86891-03-4
EINECS: 239-448-9