Benzyl-4-oxo-1-piperidinecarboxylate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine
B23181.06 B23181.14
B23181.06EA 2150 SEK
B23181.06 B23181.14
Benzyl-4-oxo-1-piperidinecarboxylate ≥98%
Benzyl-4-oxo-1-piperidinecarboxylate
Purity: 98+% CAS No.: 19099-93-5 Molecular Formula: C13H15NO3 Formula Weight: 233.27 Flash Point: 110°(230°F) Melting Point: 38-41° MDL No.: MFCD00673144
Formel: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Smältpunkt: 38…41 °C
Densitet: 1,172
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00673144
CAS nummer: 19099-93-5