2-Methylthiazolidine 98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
B22537.06
B22537.14
B22537.06EA
1325
SEK
B22537.06
B22537.14
2-Methylthiazolidine 98%
2-Methylthiazolidine
Purity: 99%
CAS No.: 24050-16-6
Molecular Formula: C4H9NS
Formula Weight: 103.18
Flash Point: 35°(95°F)
Refractive Index: 1.5260
Boiling Point: 76-78°/36mm
UN No.: UN1993
MDL No.: MFCD00053110
Formel:
C₄H₉NS Kokpunkt: 76…78 °C (36 mmHg) Flampunkt: 35 °C (95 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00053110 CAS nummer: 24050-16-6 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...