2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%
Leverantör: Thermo Fisher Scientific
Synonyms: 4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride
B22473.03 B22473.06
B22473.03EA 1950 SEK
B22473.03 B22473.06
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
Purity: 97% CAS No.: 3251-69-2 Molecular Formula: C5H6N2O2.HCl Formula Weight: 162.58 Odour: Odourless Melting Point: 221-224° Beilstein Reference: 3701591 MDL No.: MFCD00012698
Formel: C₅H₇ClN₂O₂
MW: 162.58 g/mol
Smältpunkt: 221…224 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012698
CAS nummer: 3251-69-2
EINECS: 221-840-6