tert-Butyl methyl sulphide ≥98%
Leverantör: Thermo Scientific
Fara
B22390.06 B22390.14
B22390.06EA 915 SEK
B22390.06 B22390.14
tert-Butyl methyl sulphide ≥98%
tert-Butyl methyl sulphide
Purity: 98+% CAS No.: 6163-64-0 Molecular Formula: C5H12S Formula Weight: 104.22 Flash Point: -3°(26°F) Refractive Index: 1.4400 Odour: Unpleasant Boiling Point: 101-102° UN No.: UN1993 MDL No.: MFCD00008840
Formel: C₅H₁₂S
MW: 104,21 g/mol
Kokpunkt: 101…102 °C
Densitet: 0,826
Flampunkt: –3 °C (26 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008840
CAS nummer: 6163-64-0
ADR: 3,II