Ethylene glycol monohexyl ether 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Hexyl Cellosolve, 2-(Hexyloxy)ethanol
B22385.03 B22385.06
B22385.03EA 840 SEK
B22385.03 B22385.06
Ethylene glycol monohexyl ether 99%
Ethylene glycol monohexyl ether
Purity: 99% CAS No.: 112-25-4 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 94°(201°F) Refractive Index: 1.4330 Melting Point: -42° Boiling Point: 208° Beilstein Reference: 1734691 UN No.: UN2922 MDL No.: MFCD00045997
Formel: C₈H₁₈O₂
MW: 146.23 g/mol
Kokpunkt: 208 °C
Smältpunkt: –42 °C
Densitet: 0.888
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00045997
CAS nummer: 112-25-4
EINECS: 203-951-1
ADR: 8 III