2-Hexanoylfuran 99%
Leverantör: Thermo Fisher Scientific
B22306.06 B22306.14
B22306.06EA 935 SEK
B22306.06 B22306.14
2-Hexanoylfuran 99%
2-Hexanoylfuran
Purity: 99% CAS No.: 14360-50-0 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 107°(225°F) Refractive Index: 1.4890 Odour: Characteristic Boiling Point: 125-127°/15mm Beilstein Reference: 2956 MDL No.: MFCD00037354
Formel: C₁₀H₁₄O₂
Kokpunkt: 125…127 °C (15 mmHg)
Densitet: 0,994
Flampunkt: 107 °C (225 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00037354
CAS nummer: 14360-50-0
EINECS: 238-333-0