1,7-Dioxaspiro[5.5]undecane 98%
Leverantör: Thermo Fisher Scientific
Varning
B21664.06 B21664.14 B21664.22
B21664.06EA 1250 SEK
B21664.06 B21664.14 B21664.22
1,7-Dioxaspiro[5.5]undecane 98%
1,7-Dioxaspiro[5.5]undecane
Purity: 98% CAS No.: 180-84-7 Molecular Formula: C9H16O2 Formula Weight: 156.23 Flash Point: 63°(145°F) Refractive Index: 1.4640 Boiling Point: 193° Beilstein Reference: 105997 MDL No.: MFCD00011578
Formel: C₉H₁₆O₂
Kokpunkt: 193 °C
Densitet: 1,020
Flampunkt: 63 °C (145 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011578
CAS nummer: 180-84-7
EINECS: 205-870-7