Pseudothiohydantoin 97%
Leverantör: Thermo Scientific
Synonyms: 2-aminothiazol-4(5H)-one, 2-Amino-4,5-dihydro-1,3-thiazol-4-one
B21461.06 B21461.14 B21461.22
B21461.06EA 865 SEK
B21461.06 B21461.14 B21461.22
Pseudothiohydantoin 97%
Pseudothiohydantoin
Purity: 97% CAS No.: 556-90-1 Molecular Formula: C3H4N2OS Formula Weight: 116.14 Melting Point: ca 240° dec. MDL No.: MFCD00003186
Formel: C₃H₄N₂OS
MW: 116,14 g/mol
Smältpunkt: ∼ 240 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00003186
CAS nummer: 556-90-1
EINECS: 209-145-6