Zirkonium(IV)oxid cement
Leverantör: Thermo Scientific
Varning
Synonyms: Zirconium dioxide, Zirconium Oxide
42493.30 42493.A1
42493.30EA 2575 SEK
42493.30 42493.A1
Zirkonium(IV)oxid cement
Zirconium(IV) oxide
CAS No.: 1314-23-4 Physical Form: Liquid/paste Appearance: Pale brown Odour: Odourless Merck Reference: 14,10180 MDL No.: MFCD01868884
Formel: ZrO₂
MW: 123,22 g/mol
Kokpunkt: 4300 °C (1013 hPa)
Smältpunkt: 2680 °C
Densitet: 5,75 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180